Learn More
Sigma Aldrich Fine Chemicals Biosciences Isobutyl alcohol 99 fCC fG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences W21790520KGK
Encompass
Isobutyl alcohol is a water-soluble aliphatic alcohol that can be used as a flavoring agent. It has been identified as one of the aroma/flavor compounds produced by Saccharomyces cerevisiae during the semi-aerobic fermentation of grapes to form wine.
Catalog No. 50-290-2820