Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isobutyl Benzoate 99.0+%, TCI America™
SDP

Catalog No. B0072500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0072500G 500 g
B007225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0072500G Supplier TCI America Supplier No. B0072500G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 120-50-3
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.23
MDL Number MFCD00048344
InChI Key KYZHGEFMXZOSJN-UHFFFAOYSA-N
Synonym isobutyl benzoate, benzoic acid, isobutyl ester, benzoic acid, 2-methylpropyl ester, unii-kq6xz9wjii, isobutyl benzoate natural, fema no. 2185, kq6xz9wjii, benzoic acid isobutyl ester, benzoic acid, isobutyl ester 6ci,7ci,8ci, iso-butyl benzoate
PubChem CID 61048
ChEBI CHEBI:87500
IUPAC Name 2-methylpropyl benzoate
SMILES CC(C)COC(=O)C1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 237°C
Quantity 500 g
Formula Weight 178.23
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Isobutyl Benzoate
EINECSNumber (3)-1356
RTECSNumber DH4185000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.