Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isobutyl Decanoate 98.0+%, TCI America™
SDP

Catalog No. D002125ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D002125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D002125ML Supplier TCI America Supplier No. D002125ML

Chemical Identifiers

CAS 30673-38-2
Molecular Formula C14H28O2
Molecular Weight (g/mol) 228.38
MDL Number MFCD00059259
InChI Key AXTPGRJJIGEOOY-UHFFFAOYSA-N
Synonym Decanoic Acid Isobutyl Ester
PubChem CID 121738
ChEBI CHEBI:87570
IUPAC Name 2-methylpropyl decanoate
SMILES CCCCCCCCCC(=O)OCC(C)C

Specifications

Color Yellow
Quantity 25 mL
Formula Weight 228.38
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isobutyl Decanoate
EINECSNumber (2)-0782
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.