Learn More
Medchemexpress LLC Isobutyl-deoxynyboquinone | 1430798-22-3 | 99.7% | 326.35 | C18H18N2O4 | 25MG
SDP
Catalog No. 5000236079
Encompass_Preferred
Isobutyl-deoxynyboquinone (IB-DNQ) is a small-molecule substrate selective for NAD(P)H:quinone oxidoreductase 1 (NQO1). It is used in biochemical and cellular research to study NQO1-mediated bioactivation, reactive oxygen species generation, and enzyme-dependent cytotoxicity relevant to anticancer investigations. The compound is supplied at high reported purity and is intended for research use only.
  • Selective substrate for NQO1.
  • Facilitates study of reactive oxygen species generation.
  • Enables investigation of NQO1-dependent cytotoxicity in cells.
  • High reported purity for biochemical assays.
  • Stable when stored under recommended conditions.
Catalog No. 50-002-36079 Supplier Medchemexpress LLC Supplier No. HY12502725MG
Isobutyl-deoxynyboquinone (IB-DNQ) is a small-molecule substrate selective for NAD(P)H:quinone oxidoreductase 1 (NQO1). It is used in biochemical and cellular research to study NQO1-mediated bioactivation, reactive oxygen species generation, and enzyme-dependent cytotoxicity relevant to anticancer investigations. The compound is supplied at high reported purity and is intended for research use only.
  • Selective substrate for NQO1.
  • Facilitates study of reactive oxygen species generation.
  • Enables investigation of NQO1-dependent cytotoxicity in cells.
  • High reported purity for biochemical assays.
  • Stable when stored under recommended conditions.