Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isobutyl-deoxynyboquinone | 1430798-22-3 | 99.7% | 326.35 | C18H18N2O4 | 5MG
SDP

Catalog No. 5000235204
Encompass_Preferred

Isobutyl-deoxynyboquinone (IB-DNQ) is a small-molecule quinone used as a selective substrate for NAD(P)H:quinone oxidoreductase 1 (NQO1) in preclinical research. It is employed to probe NQO1-dependent cytotoxicity and has demonstrated in vitro potency across cancer cell lines, with documented in vivo exposure in pharmacokinetic studies.

  • Selective substrate for NQO1.
  • Used to study NQO1-mediated cytotoxicity in cell-based assays.
  • Demonstrates in vitro activity in SCCF cell lines.
  • Molecular formula C18H18N2O4; molecular weight 326.35.
  • Purity 99.7%.
  • Appearance brown to red solid.
  • Solubility DMSO 10 mg/mL; may require warming or sonication.
  • Powder storage: -20°C (up to 3 years) or 4°C (up to 2 years); stock solutions: -80°C (6 months).

Catalog No. 50-002-35204 Supplier Medchemexpress LLC Supplier No. HY1250275MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Isobutyl-deoxynyboquinone (IB-DNQ) is a small-molecule quinone used as a selective substrate for NAD(P)H:quinone oxidoreductase 1 (NQO1) in preclinical research. It is employed to probe NQO1-dependent cytotoxicity and has demonstrated in vitro potency across cancer cell lines, with documented in vivo exposure in pharmacokinetic studies.

  • Selective substrate for NQO1.
  • Used to study NQO1-mediated cytotoxicity in cell-based assays.
  • Demonstrates in vitro activity in SCCF cell lines.
  • Molecular formula C18H18N2O4; molecular weight 326.35.
  • Purity 99.7%.
  • Appearance brown to red solid.
  • Solubility DMSO 10 mg/mL; may require warming or sonication.
  • Powder storage: -20°C (up to 3 years) or 4°C (up to 2 years); stock solutions: -80°C (6 months).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.