Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isobutyl-deoxynyboquinone | 1430798-22-3 | 99.7% | 326.35 g/mol | C18H18N2O4 | 10MG
SDP

Catalog No. 5000235654
Encompass_Preferred

Isobutyl-deoxynyboquinone (IB-DNQ) is a research-grade small molecule that functions as a selective substrate for NAD(P)H:quinone oxidoreductase (NQO1). It is used in biochemical and anticancer research to probe NQO1-mediated redox activity and to induce cytotoxicity in NQO1-expressing cells. The compound is supplied as a high-purity solid and is typically dissolved in DMSO for in vitro experiments.

  • Selective substrate for NAD(P)H:quinone oxidoreductase (NQO1).
  • High purity: 99.7%.
  • Chemical formula: C18H18N2O4.
  • Molecular weight: 326.35 g/mol.
  • Soluble in DMSO at 10 mg/mL with ultrasonic and warming; heat to 60°C improves solubility.
  • Solid appearance, brown to red; store powder at -20°C for long-term stability.

Catalog No. 50-002-35654 Supplier Medchemexpress LLC Supplier No. HY12502710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isobutyl-deoxynyboquinone (IB-DNQ) is a research-grade small molecule that functions as a selective substrate for NAD(P)H:quinone oxidoreductase (NQO1). It is used in biochemical and anticancer research to probe NQO1-mediated redox activity and to induce cytotoxicity in NQO1-expressing cells. The compound is supplied as a high-purity solid and is typically dissolved in DMSO for in vitro experiments.

  • Selective substrate for NAD(P)H:quinone oxidoreductase (NQO1).
  • High purity: 99.7%.
  • Chemical formula: C18H18N2O4.
  • Molecular weight: 326.35 g/mol.
  • Soluble in DMSO at 10 mg/mL with ultrasonic and warming; heat to 60°C improves solubility.
  • Solid appearance, brown to red; store powder at -20°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.