Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isobutyl Gallate 98.0+%, TCI America™
SDP

Catalog No. G02055G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G02055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G02055G Supplier TCI America Supplier No. G02055G
Add to Cart
Add to Cart

Specifications

CAS 3856-5-1
Melting Point 132°C
Color White
Molecular Formula C11H14O5
Quantity 5 g
MDL Number MFCD00016425
Synonym Gallic Acid Isobutyl Ester
InChI Key UCLHVCFKZSLALE-UHFFFAOYSA-N
SMILES CC(C)COC(=O)C1=CC(=C(C(=C1)O)O)O
IUPAC Name 2-methylpropyl 3,4,5-trihydroxybenzoate
Molecular Weight (g/mol) 226.23
PubChem CID 77464
Formula Weight 226.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Isobutyl Gallate
Show More Show Less
EINECSNumber (3)-1583
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.