Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger TCI AMERICA Isobutyl Methacrylate, Reagent, >98.0% GC, 25 mL

Catalog No. 502780953
Encompass_Preferred

Isobutyl methacrylate is a chemical substance supplied for laboratory research purposes. This reagent is stabilized with hydroquinone to maintain product integrity and is provided in a clear glass bottle. It is classified as a flammable liquid and requires careful handling in accordance with standard safety protocols.

  • Formula: C8H14O2
  • Purity: >98.0% GC
  • Molecular weight: 142.20
  • Boiling point: 155°C (311°F)
  • Flash point: 42°C (108°F)
  • CAS Number: 97-86-9
  • Relative density: 0.89
  • Stabilizer: Hydroquinone

Catalog No. 50-278-0953 Supplier Grainger Supplier No. 19KT82
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isobutyl methacrylate is a chemical substance supplied for laboratory research purposes. This reagent is stabilized with hydroquinone to maintain product integrity and is provided in a clear glass bottle. It is classified as a flammable liquid and requires careful handling in accordance with standard safety protocols.

  • Formula: C8H14O2
  • Purity: >98.0% GC
  • Molecular weight: 142.20
  • Boiling point: 155°C (311°F)
  • Flash point: 42°C (108°F)
  • CAS Number: 97-86-9
  • Relative density: 0.89
  • Stabilizer: Hydroquinone

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.