Learn More
Isobutyl methacrylate, 99.5+%, stab. with 10ppm 4-methoxyphenol
Description
Plastic industry: used in the manufacture of containing active hydroxyl acrylic resin. Coatings with epoxy resin, diisocyanate and melamine formaldehyde resin configuration used for preparing a two-component coating. In oil industry as additives for lubricating oil washing. In electronics industry used as electron microscope to take off the water. Textile industry used in the manufacture of adhesive fabric. Used as chemical reagents in analytical chemistry. In addition, also used for water immiscible embedding agent, used in the synthesis of medical polymer materials, thermosetting coatings and adhesives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 0.889 g/mL |
| Boiling Point | 155°C |
| Flash Point | 46°C (115°F) |
| Quantity | 1 L |
| UN Number | UN2283 |
| Beilstein | 1747595 |
| Solubility Information | Soluble in water at 20°C,2g/L |
| Formula Weight | 142.2 |
| Percent Purity | ≥99.5% |
| Physical Form | Liquid |
| Show More |
Safety and Handling
| Hazard Category | H226-H315-H317-H319-H335 |
| Hazard Statement | GHS H Statement H226-H315-H319-H317-H335 Flammable liquid and vapor. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P333+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ISOBUTYL METHACRYLATE, STABILIZED |
| EINECSNumber | 202-613-0 |
| RTECSNumber | OZ4900000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.