Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isobutyl Methanesulfonate 98.0+%, TCI America™
SDP

Catalog No. I10621G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I10621G 1 g
I10625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I10621G Supplier TCI America Supplier No. I10621G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16156-53-9
Molecular Formula C5H12O3S
Molecular Weight (g/mol) 152.208
MDL Number MFCD07363982
InChI Key PZMKMCFRTKGVOL-UHFFFAOYSA-N
Synonym Methanesulfonic Acid Isobutyl Ester
PubChem CID 119240
IUPAC Name 2-methylpropyl methanesulfonate
SMILES CC(C)COS(=O)(=O)C

Specifications

Color Yellow
Boiling Point 53°C
Quantity 1 g
Formula Weight 152.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isobutyl Methanesulfonate
RTECSNumber PB2662500
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.