Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Isobutyl nitrite

Catalog No. 501857656
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-185-7656 100 mL
50-185-7657 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-185-7656 Supplier Sigma Aldrich Supplier No. 327190100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Isobutyl nitrite was extensively used as room odorizers
  • It is a popular recreational drug.isobutyl nitrite was used to study inhalant-induced immunotoxicity
TRUSTED_SUSTAINABILITY

Specifications

CAS 542-56-3
Density 0.87 g/mL (at 25°C (literature))
Boiling Point 66°C to 67°C (lit.)
Molecular Formula C4H9NO2
Refractive Index n20/D 1.373 (literature)
Linear Formula (CH3)2 CHCH2ONO
MDL Number MFCD00002056
Quantity 100 mL
Molecular Weight (g/mol) 103.12
Percent Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.