Learn More
Medchemexpress LLC Isobutyl p-hydroxybenzoate | 4247-02-3 | MFCD00020167 | 98.9% | 194.23 g/mol | C11H14O3 | 1 KG
SDP
Supplier:  Medchemexpress LLC HYW0150261KG
Encompass_Preferred
Isobutyl p-hydroxybenzoate (isobutylparaben) is a benzoate ester used as a laboratory reagent and reported agonist of nuclear receptors including PXR, CAR, and PPAR. It has broad-spectrum antimicrobial properties and is used in studies of receptor activation, toxicology, and formulation development. Typical handling uses DMSO as solvent and the material is supplied at high purity appropriate for biochemical assays.
  • Agonist of PXR, CAR, and PPAR nuclear receptors.
  • Broad-spectrum antimicrobial preservative properties.
  • High purity suitable for biochemical and toxicology studies.
  • Soluble in DMSO for preparation of stock solutions.
  • Available in laboratory and bulk pack sizes for various research needs.
Catalog No. 50-003-03353