Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isobutylamido thiazolyl resorcinol | 1428450-95-6 | MFCD34165742 | 99.9% | 278.33 | C13H14N2O3S | 10 G
SDP

Catalog No. 5000281701
Encompass_Preferred

Isobutylamido thiazolyl resorcinol is a small-molecule tyrosinase inhibitor used in research to prevent UV-induced pigmentation. Supplied as a solid for laboratory use, the compound includes manufacturer guidance for storage and handling and provides solubility and formulation options for in vitro and in vivo studies.

  • Tyrosinase inhibitor that reduces UV-induced pigmentation.
  • Solid form suitable for standard laboratory handling.
  • High purity (99.9%).
  • Molecular weight 278.33 g/mol.
  • Soluble in DMSO at 100 mg/mL; in vivo vehicles achieve ≥5 mg/mL.
  • Storage and handling information provided for refrigerated and solvent conditions.

Catalog No. 50-002-81701 Supplier Medchemexpress LLC Supplier No. HYW25118110G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isobutylamido thiazolyl resorcinol is a small-molecule tyrosinase inhibitor used in research to prevent UV-induced pigmentation. Supplied as a solid for laboratory use, the compound includes manufacturer guidance for storage and handling and provides solubility and formulation options for in vitro and in vivo studies.

  • Tyrosinase inhibitor that reduces UV-induced pigmentation.
  • Solid form suitable for standard laboratory handling.
  • High purity (99.9%).
  • Molecular weight 278.33 g/mol.
  • Soluble in DMSO at 100 mg/mL; in vivo vehicles achieve ≥5 mg/mL.
  • Storage and handling information provided for refrigerated and solvent conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.