Learn More
Medchemexpress LLC Isobutylamido thiazolyl resorcinol | 1428450-95-6 | MFCD34165742 | 99.9% | 278.33 g/mol | C13H14N2O3S | 1 G
SDP
Catalog No. 5000281246
Encompass_Preferred
Isobutylamido thiazolyl resorcinol is a small-molecule tyrosinase inhibitor used in research to investigate melanin synthesis and evaluate depigmenting strategies. It is supplied as a white to off-white solid with a molecular weight of 278.33 g/mol and high analytical purity, and is commonly applied in topical formulation testing and cellular assays.
  • High purity suitable for research applications.
  • White to off-white solid for straightforward handling.
  • Potent tyrosinase inhibition for pigmentation and melanin studies.
  • Multiple packaging options for dose flexibility.
  • Available as a ready-to-use 10 mM solution in DMSO for assays.
Catalog No. 50-002-81246 Supplier Medchemexpress LLC Supplier No. HYW2511811G
Isobutylamido thiazolyl resorcinol is a small-molecule tyrosinase inhibitor used in research to investigate melanin synthesis and evaluate depigmenting strategies. It is supplied as a white to off-white solid with a molecular weight of 278.33 g/mol and high analytical purity, and is commonly applied in topical formulation testing and cellular assays.
  • High purity suitable for research applications.
  • White to off-white solid for straightforward handling.
  • Potent tyrosinase inhibition for pigmentation and melanin studies.
  • Multiple packaging options for dose flexibility.
  • Available as a ready-to-use 10 mM solution in DMSO for assays.