Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isobutylamido thiazolyl resorcinol | 1428450-95-6 | MFCD34165742 | 99.9% | 278.33 g/mol | C13H14N2O3S | 1 ML
SDP

Catalog No. 5000281247
Encompass_Preferred

Isobutylamido thiazolyl resorcinol is a research-grade small-molecule tyrosinase inhibitor used in studies of pigmentation. It is supplied as a high-purity solid and as a ready-to-use 10 mM solution in DMSO for laboratory research; intended for research use only.

  • High purity (99.88%).
  • Available as 10 mM solution in DMSO (1 mL) and as solids (500 mg-25 g).
  • Targets tyrosinase and can prevent UV-induced pigmentation.
  • Molecular formula C13H14N2O3S; molecular weight 278.33 g/mol.
  • Provided with manufacturer datasheet for research handling information.

Catalog No. 50-002-81247 Supplier Medchemexpress LLC Supplier No. HYW25118110MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isobutylamido thiazolyl resorcinol is a research-grade small-molecule tyrosinase inhibitor used in studies of pigmentation. It is supplied as a high-purity solid and as a ready-to-use 10 mM solution in DMSO for laboratory research; intended for research use only.

  • High purity (99.88%).
  • Available as 10 mM solution in DMSO (1 mL) and as solids (500 mg-25 g).
  • Targets tyrosinase and can prevent UV-induced pigmentation.
  • Molecular formula C13H14N2O3S; molecular weight 278.33 g/mol.
  • Provided with manufacturer datasheet for research handling information.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.