Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isobutylamido thiazolyl resorcinol | 1428450-95-6 | MFCD34165742 | 99.9% | 278.33 g·mol⁻¹ | C13H14N2O3S | 25 G
SDP

Catalog No. 5000280645
Encompass_Preferred

Isobutylamido thiazolyl resorcinol is a small-molecule tyrosinase inhibitor used in research to prevent UV-induced pigmentation. Supplied as a high-purity solid for laboratory studies, it is intended for in vitro assays, melanogenesis research, and compound screening. Consult the safety data sheet for handling and storage instructions.

  • 99.9% purity
  • Molecular formula C13H14N2O3S
  • Molecular weight 278.33 g·mol⁻¹
  • Tyrosinase inhibitor that prevents UV-induced pigmentation
  • Available as a 25 G solid
  • Store under nitrogen; in solvent: -80°C for 6 months, -20°C for 1 month
  • For research use only; not for human or veterinary use

Catalog No. 50-002-80645 Supplier Medchemexpress LLC Supplier No. HYW25118125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isobutylamido thiazolyl resorcinol is a small-molecule tyrosinase inhibitor used in research to prevent UV-induced pigmentation. Supplied as a high-purity solid for laboratory studies, it is intended for in vitro assays, melanogenesis research, and compound screening. Consult the safety data sheet for handling and storage instructions.

  • 99.9% purity
  • Molecular formula C13H14N2O3S
  • Molecular weight 278.33 g·mol⁻¹
  • Tyrosinase inhibitor that prevents UV-induced pigmentation
  • Available as a 25 G solid
  • Store under nitrogen; in solvent: -80°C for 6 months, -20°C for 1 month
  • For research use only; not for human or veterinary use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.