Learn More
Medchemexpress LLC Isobutylamido thiazolyl resorcinol | 1428450-95-6 | MFCD34165742 | 99.9% | 278.33 g/mol | C13H14N2O3S | 5 G
SDP
Catalog No. 5000281539
Encompass_Preferred
Isobutylamido thiazolyl resorcinol is a small-molecule tyrosinase inhibitor used in laboratory research to prevent UV-induced pigmentation and to study melanin synthesis pathways. The compound is supplied as a solid and as a DMSO solution, with high reported purity and documented storage recommendations to preserve stability. For research use only.
  • Potent tyrosinase inhibition for pigmentation studies.
  • High purity (~99.9%) suitable for analytical and biological assays.
  • Available as solid and DMSO solution formats for experimental flexibility.
  • Documented storage conditions to maintain compound integrity.
  • Safety data sheet and certificate of analysis available for quality and handling information.
  • Suitable for in vitro studies of melanin synthesis and related pathways.
Catalog No. 50-002-81539 Supplier Medchemexpress LLC Supplier No. HYW2511815G
Isobutylamido thiazolyl resorcinol is a small-molecule tyrosinase inhibitor used in laboratory research to prevent UV-induced pigmentation and to study melanin synthesis pathways. The compound is supplied as a solid and as a DMSO solution, with high reported purity and documented storage recommendations to preserve stability. For research use only.
  • Potent tyrosinase inhibition for pigmentation studies.
  • High purity (~99.9%) suitable for analytical and biological assays.
  • Available as solid and DMSO solution formats for experimental flexibility.
  • Documented storage conditions to maintain compound integrity.
  • Safety data sheet and certificate of analysis available for quality and handling information.
  • Suitable for in vitro studies of melanin synthesis and related pathways.