Learn More
Medchemexpress LLC Isobutylamido thiazolyl resorcinol | 1428450-95-6 | MFCD34165742 | 99.9% | 278.33 g/mol | C13H14N2O3S | 500 MG
SDP
Catalog No. 5000280667
Encompass_Preferred
Isobutylamido thiazolyl resorcinol is a small-molecule tyrosinase inhibitor used in laboratory research to study and prevent UV-induced pigmentation. It is supplied as a high-purity solid suitable for biochemical and cellular assays and is intended for research use only.
  • Tyrosinase inhibitor for pigmentation research.
  • High purity (99.9%) suitable for analytical and biological studies.
  • Molecular formula C13H14N2O3S and molecular weight 278.33 g/mol.
  • Supplied as a 500 mg solid pack for laboratory use.
  • Shipped with safety data sheet and product documentation.
  • Not for human or veterinary use; for research use only.
Catalog No. 50-002-80667 Supplier Medchemexpress LLC Supplier No. HYW251181500MG
Isobutylamido thiazolyl resorcinol is a small-molecule tyrosinase inhibitor used in laboratory research to study and prevent UV-induced pigmentation. It is supplied as a high-purity solid suitable for biochemical and cellular assays and is intended for research use only.
  • Tyrosinase inhibitor for pigmentation research.
  • High purity (99.9%) suitable for analytical and biological studies.
  • Molecular formula C13H14N2O3S and molecular weight 278.33 g/mol.
  • Supplied as a 500 mg solid pack for laboratory use.
  • Shipped with safety data sheet and product documentation.
  • Not for human or veterinary use; for research use only.