Learn More
Accela Chembio Inc Isobutylboronic Acid | 5g | 84110-40-7 | MFCD00134156 | 95+% | Shelf Life: 1260 Days | Moisture Sensitive/nitrogen Or Argon/+4
Catalog No. 50249820
Encompass_Preferred
Isobutylboronic Acid | 5g | 84110-40-7 | MFCD00134156 | 95+% | Shelf Life: 1260 Days | Moisture Sensitive/nitrogen Or Argon/+4
Catalog No. 50-249-820 Supplier Accela Chembio Inc Supplier No. SY0024415G
Isobutylboronic Acid | 5g | 84110-40-7 | MFCD00134156 | 95+% | Shelf Life: 1260 Days | Moisture Sensitive/nitrogen Or Argon/+4