Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isobutylcyclohexane 98.0+%, TCI America™
SDP

Catalog No. I025925ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I025925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I025925ML Supplier TCI America Supplier No. I025925ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1678-98-4
Molecular Formula C10H20
Molecular Weight (g/mol) 140.27
MDL Number MFCD00013767
InChI Key FFROMNOQCNVNIH-UHFFFAOYSA-N
Synonym isobutylcyclohexane, cyclohexane, isobutyl, cyclohexane, 2-methylpropyl, i-butylcyclohexane, acmc-209dx9, 2-methyl-propyl-cyclohexane
PubChem CID 15508
IUPAC Name 2-methylpropylcyclohexane
SMILES CC(C)CC1CCCCC1

Specifications

Color Colorless
Boiling Point 169°C
Quantity 25 mL
UN Number 3295
Formula Weight 140.27
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isobutylcyclohexane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.