Learn More
RAE Systems Inc ISOBUTYLENE CAL GAS 34L
SDP
Catalog No. NC9146106
Encompass_Preferred
This item has a minimum qty of 5 per supplier requirements.
Isobutylene, 100 ppm (balance air), 34L steel cylinder
Catalog No. NC9146106 Supplier RAE Systems Inc Supplier No. 6000002000
This item has a minimum qty of 5 per supplier requirements.
Isobutylene, 100 ppm (balance air), 34L steel cylinder