Learn More
Medchemexpress LLC Isobutylparaben-d4 | 1219805-33-0 | 98.5% | 198.25 g/mol | C11H10D4O3 | 1 MG
SDP
Catalog No. 5000302517
Encompass_Preferred
Isobutylparaben-d4 is the deuterium-labeled analogue of isobutylparaben used as a stable isotope internal standard for analytical and research applications. It is provided as a high-purity, fully characterized material designed to support quantitative mass spectrometry, method development, and calibration workflows.
  • High isotopic enrichment for accurate internal standardization.
  • High purity suitable for quantitative analytical use.
  • Optimized for mass spectrometry and chromatographic methods.
  • Supplied in measured small quantities for standards preparation.
  • Certificate of analysis documents purity and isotopic enrichment.
Catalog No. 50-003-02517 Supplier Medchemexpress LLC Supplier No. HYW015026S1MG
Isobutylparaben-d4 is the deuterium-labeled analogue of isobutylparaben used as a stable isotope internal standard for analytical and research applications. It is provided as a high-purity, fully characterized material designed to support quantitative mass spectrometry, method development, and calibration workflows.
  • High isotopic enrichment for accurate internal standardization.
  • High purity suitable for quantitative analytical use.
  • Optimized for mass spectrometry and chromatographic methods.
  • Supplied in measured small quantities for standards preparation.
  • Certificate of analysis documents purity and isotopic enrichment.