Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isobutylparaben-d4 (isobutyl 4-hydroxybenzoate-d4) | 1219805-33-0 | 98.5% | 198.25 g/mol | C11H10D4O3 | 5 MG
SDP

Catalog No. 5000302267
Encompass_Preferred

Isobutylparaben-d4 is the deuterium-labeled analogue of isobutylparaben used as an analytical standard for quantitative assays, including LC-MS, where isotope dilution improves accuracy and precision. Supplied as a purified powder for research use, it facilitates trace-level detection and method validation.

  • Deuterium-labeled internal standard for LC-MS quantitation.
  • High purity for accurate analytical results.
  • Molecular weight 198.25 g/mol and formula C11H10D4O3.
  • Powder form with recommended storage at -20°C; solvent stability at -80°C (6 months) and -20°C (1 month).
  • Ideal small-quantity packaging for method development and validation.

Catalog No. 50-003-02267 Supplier Medchemexpress LLC Supplier No. HYW015026S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Isobutylparaben-d4 is the deuterium-labeled analogue of isobutylparaben used as an analytical standard for quantitative assays, including LC-MS, where isotope dilution improves accuracy and precision. Supplied as a purified powder for research use, it facilitates trace-level detection and method validation.

  • Deuterium-labeled internal standard for LC-MS quantitation.
  • High purity for accurate analytical results.
  • Molecular weight 198.25 g/mol and formula C11H10D4O3.
  • Powder form with recommended storage at -20°C; solvent stability at -80°C (6 months) and -20°C (1 month).
  • Ideal small-quantity packaging for method development and validation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.