Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isobutyltrichlorosilane 98.0+%, TCI America™
SDP

Catalog No. I0452500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0452500G 500 g
I045225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0452500G Supplier TCI America Supplier No. I0452500G

Chemical Identifiers

CAS 18169-57-8
Molecular Formula C4H9Cl3Si
Molecular Weight (g/mol) 191.551
MDL Number MFCD00053185
InChI Key GBXOGFTVYQSOID-UHFFFAOYSA-N
Synonym isobutyltrichlorosilane, trichloroisobutylsilane, silane, trichloro 2-methylpropyl, trichloro isobutyl silane, trichloro 2-methylpropyl silane, i-butyltrichlorosilane, silane,trichloro 2-methylpropyl, acmc-209ei4, trichloro isobutyl silane #, gbxogftvyqsoid-uhfffaoysa
PubChem CID 87489
IUPAC Name trichloro(2-methylpropyl)silane
SMILES CC(C)C[Si](Cl)(Cl)Cl

Specifications

Color Colorless
Boiling Point 140°C
Quantity 500 g
UN Number 1747
Formula Weight 191.55
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isobutyltrichlorosilane
EINECSNumber (2)-2041
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.