Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isobutyltriphenylphosphonium bromide, 98+%, Thermo Scientific™

Catalog No. AAA1788122
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1788122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1788122 Supplier Thermo Scientific Chemicals Supplier No. A1788122

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is employed as intermediate for pharmaceutical.

Notes
Hygroscopic, store away from moisture. Store in a tightly closed container in a cool, dry, well-ventilated area. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22884-29-3
Molecular Formula C22H24BrP
Molecular Weight (g/mol) 399.31
MDL Number MFCD00031593
InChI Key VIKPDPHNYHUZQW-UHFFFAOYSA-M
Synonym isobutyltriphenylphosphonium bromide, 2-methylpropyl triphenyl phosphanium bromide, phosphonium, 2-methylpropyl triphenyl-, bromide, 2-methylpropyl triphenylphosphanium bromide, acmc-1citv, isobutyltriphenylphosphoniumbromide, isobutyl triphenylphosphonium bromide, 2-methylpropyl triphenyl phosphonium bromide, isobutyl triphenyl phosphonium bromide, phosphonium, 2-methylpropyl triphenyl-,bromide 1:1
PubChem CID 2724571
SMILES [Br-].CC(C)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 198°C to 202°C
Quantity 100 g
Beilstein 4171228
IUPAC Name (2-methylpropyl)triphenylphosphanium bromide
Formula Weight 399.32
Percent Purity ≥98%
Chemical Name or Material Isobutyltriphenylphosphonium bromide
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 245-291-7
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.