Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isobutyltriphenylphosphonium bromide, 99%, Thermo Scientific™

Catalog No. AC259280500
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC259280500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC259280500 Supplier Thermo Scientific Chemicals Supplier No. 259280500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22884-29-3
Molecular Formula C22H24BrP
Molecular Weight (g/mol) 399.31
MDL Number MFCD00031593
InChI Key VIKPDPHNYHUZQW-UHFFFAOYSA-M
Synonym isobutyltriphenylphosphonium bromide, 2-methylpropyl triphenyl phosphanium bromide, phosphonium, 2-methylpropyl triphenyl-, bromide, 2-methylpropyl triphenylphosphanium bromide, acmc-1citv, isobutyltriphenylphosphoniumbromide, isobutyl triphenylphosphonium bromide, 2-methylpropyl triphenyl phosphonium bromide, isobutyl triphenyl phosphonium bromide, phosphonium, 2-methylpropyl triphenyl-,bromide 1:1
PubChem CID 2724571
IUPAC Name (2-methylpropyl)triphenylphosphanium bromide
SMILES [Br-].CC(C)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 200°C to 203°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Linear Formula [(C6H5)3-P-i-C4H9]Br
Quantity 50 g
Formula Weight 399.29
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material Isobutyltriphenylphosphonium bromide, 99%
Hazard Category LIGHT SENSITIVE
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.