Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Isobutylzinc Bromide Solution

Catalog No. 49896350ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-896-350ML 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 49-896-350ML Supplier Sigma Aldrich Supplier No. 49896350ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Isobutylzinc bromide is an organozinc compound, which can be used as a reagent in the palladium-catalyzed Negishi cross-coupling reaction to construct a carbon-carbon bond by reacting with aryl or heteroaryl halides . It is also used to prepare regioselective unsaturated ketones by reacting with tert-butyl isocyanide via an allylic carbonylative coupling reaction in the presence of a nickel catalyst.Product of Rieke Metals, Inc. Rieke is a registered trademark of Rieke Metals, Inc.

Specifications

CAS 45438-80-0
Density 0.973 g/mL at 25°C
MDL Number MFCD00283488
Quantity 5 g
Synonym (Hydroxymethyl)polystyrene; Benzyl alcohol polymer-bound; Hydroxymethyl polystyrene; Polystyrene cross-linked with divinylbenzene, hydroxymethylated
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.