Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Isobutyraldehyde

Catalog No. 418110800ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
800 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-811-0800ML 800 mL
41-811-0100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 41-811-0800ML Supplier Sigma Aldrich Supplier No. 418110800ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Isobutyraldehyde

Specifications

CAS 78-84-2
Density 0.79 g/mL (at 25°C (literature))
Molecular Formula C4H8O
Linear Formula (CH3)2 CHCHO
MDL Number MFCD00006980
Melting Point -65°C (lit.)
Boiling Point 63°C (lit.)
Synonym 2-Methylpropanal; 2-Methylpropionaldehyde
Molecular Weight (g/mol) 72.11
Quantity 800 mL
Percent Purity ≥99.5%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.