Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Isobutyric anhydride

Catalog No. 245771500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-577-1500ML 500 mL
24-577-12L 2 L
24-577-125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 24-577-1500ML Supplier Sigma Aldrich Supplier No. 245771500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Isobutyric anhydride

Specifications

Melting Point -56°C (lit.)
CAS 97-72-3
Boiling Point 182°C (lit.)
Density 0.954 g/mL (at 25°C (literature))
Quantity 500 mL
Refractive Index n20/D 1.406 (literature)
Molecular Formula C8H14O3
Linear Formula (CH3)2 CHCO2COCH(CH3)2
MDL Number MFCD00008913
Synonym 2-Methylpropionic anhydride
Molecular Weight (g/mol) 158.19
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.