Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chem-Impex International, Inc. Isobutyric anhydride, distilled | 97-72-3 | MFCD00008913 | 5L

Catalog No. 50492121
Encompass_Preferred

Isobutyric anhydride, distilled, 97-72-3, MFCD00008913, 5L

Catalog No. 50-492-121 Supplier Chem-Impex International, Inc. Supplier No. 007065L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isobutyric anhydride, distilled, 97-72-3, MFCD00008913, 5L

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.