Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aobchem Isobutyrophenone | ≥95% | CAS 611-70-1 | C10H12O | 100g

Catalog No. 5001043160
Encompass_Preferred

Isobutyrophenone | ≥95% | CAS 611-70-1 | C10H12O | 100g

Catalog No. 50-010-43160 Supplier Aobchem Supplier No. A339178100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isobutyrophenone | ≥95% | CAS 611-70-1 | C10H12O | 100g

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.