Learn More
Cayman Chemical Isobutyryl-L-carnitine-d3 (chloride), Purity: ≥99% deuterated forms (d1-d3), 5 mg
SDP
Catalog No. 502935121
Encompass_Preferred
An internal standard for the quantification of isobutyryl-L-carnitine by GC- or LC-MS
Catalog No. 50-293-5121 Supplier Cayman Chemical Supplier No. 265765 MG
An internal standard for the quantification of isobutyryl-L-carnitine by GC- or LC-MS