Learn More
Medchemexpress LLC Isobutyryl-L-carnitine | 25518-49-4 | MFCD01674829 | 99.9% | 231.29 g/mol | C11H21NO4 | 5 MG
SDP
Catalog No. 5000263540
Encompass_Preferred
Isobutyryl-L-carnitine is an endogenous acylcarnitine provided as a solid analytical standard for biochemical and metabolomics research. It serves as a reference compound for studies of fatty acid oxidation, acylcarnitine profiling, and diagnostic assays.
  • High purity suitable for analytical use.
  • Supplied as a white to off-white solid.
  • Molecular weight 231.29 g/mol for accurate mass reference.
  • Available in small research quantities and solution formats.
  • Intended for metabolomics and biochemical reference applications.
Catalog No. 50-002-63540 Supplier Medchemexpress LLC Supplier No. HY1131655MG
Isobutyryl-L-carnitine is an endogenous acylcarnitine provided as a solid analytical standard for biochemical and metabolomics research. It serves as a reference compound for studies of fatty acid oxidation, acylcarnitine profiling, and diagnostic assays.
  • High purity suitable for analytical use.
  • Supplied as a white to off-white solid.
  • Molecular weight 231.29 g/mol for accurate mass reference.
  • Available in small research quantities and solution formats.
  • Intended for metabolomics and biochemical reference applications.