Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isobutyryl-L-carnitine chloride | 6920-31-6 | MFCD01674829 | ≥95.0% | 267.75 | C11H22ClNO4 | 5 MG
SDP

Catalog No. 5000264793
Encompass_Preferred

Isobutyryl-L-carnitine chloride is an endogenous acylcarnitine provided as the chloride salt for laboratory research. It is used in biochemical, metabolic, and analytical studies related to fatty acid oxidation and acyl-CoA dehydrogenase pathways, and is supplied in small quantities suitable for analytical assays.

  • Endogenous metabolite used as a biochemical reagent.
  • Supplied as the chloride salt for improved stability and handling.
  • Molecular formula C11H22ClNO4; molecular weight 267.75.
  • CAS number 6920-31-6 for unambiguous identification.
  • Available in small masses (for example, 5 mg) for analytical and prep work.
  • For research use only; not for human or clinical applications.

Catalog No. 50-002-64793 Supplier Medchemexpress LLC Supplier No. HY113165A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isobutyryl-L-carnitine chloride is an endogenous acylcarnitine provided as the chloride salt for laboratory research. It is used in biochemical, metabolic, and analytical studies related to fatty acid oxidation and acyl-CoA dehydrogenase pathways, and is supplied in small quantities suitable for analytical assays.

  • Endogenous metabolite used as a biochemical reagent.
  • Supplied as the chloride salt for improved stability and handling.
  • Molecular formula C11H22ClNO4; molecular weight 267.75.
  • CAS number 6920-31-6 for unambiguous identification.
  • Available in small masses (for example, 5 mg) for analytical and prep work.
  • For research use only; not for human or clinical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.