Learn More
Medchemexpress LLC Isobutyryl-L-carnitine-d3 chloride | 6920-31-6 | 99.5% | 270.77 | 1 MG
SDP
Catalog No. 5000437227
Encompass_Preferred
Isobutyryl-L-carnitine-d3 (chloride) is the deuterium labeled Isobutyryl-L-carnitine chloride. Isobutyryl-L-carnitine chloride is a product of the acyl-CoA dehydrogenases.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-37227 Supplier Medchemexpress LLC Supplier No. HY113165AS1MG
Isobutyryl-L-carnitine-d3 (chloride) is the deuterium labeled Isobutyryl-L-carnitine chloride. Isobutyryl-L-carnitine chloride is a product of the acyl-CoA dehydrogenases.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs