Learn More
Cayman Chemical IsobutyrylCoenzyme AsodIM 10mg
SDP
Catalog No. 502941673
Encompass_Preferred
A short-chain branched acyl CoA; a substrate for IBDH
Catalog No. 50-294-1673 Supplier Cayman Chemical Supplier No. 2786710 MG
A short-chain branched acyl CoA; a substrate for IBDH