Learn More
Cayman Chemical Isobutyryl-L-carnitine (chloride), Purity: ≥95%, CAS: 6920-31-6, 50 mg
SDP
Catalog No. 502915035
Encompass_Preferred
An acylcarnitine; elevated levels are associated with isobutyryl-CoA dehydrogenase deficiency
Catalog No. 50-291-5035 Supplier Cayman Chemical Supplier No. 3710350 MG
An acylcarnitine; elevated levels are associated with isobutyryl-CoA dehydrogenase deficiency