Learn More
MSC Dupont IsoClean Hood, Tyvek 400, Bound Seams, Medium, 100/Pk.
Catalog No. 5000684193
Encompass
This clean and sterile hood is designed for use in controlled environments and critical applications, including pharmaceutical, biotechnology, and medical device manufacturing. Constructed from flash-spun polyethylene fabric, it offers a robust barrier against particles, microorganisms, and non-hazardous liquids. The hood is processed and individually packaged in an ISO Class 4 cleanroom to minimize particle shedding and ensure high garment cleanliness.
  • Provides an excellent barrier to particles, microorganisms, and non-hazardous liquids
  • Suitable for ISO Classes 4-9 cleanrooms
  • Constructed with bound seams for enhanced protection
  • Treated with a topical antistatic agent
  • Gamma irradiated to achieve a sterility assurance level of 10-6
  • Free of natural rubber latex
  • Individually packaged for convenience in controlled settings
Catalog No. 50-006-84193 Supplier MSC Supplier No. 44629897
This clean and sterile hood is designed for use in controlled environments and critical applications, including pharmaceutical, biotechnology, and medical device manufacturing. Constructed from flash-spun polyethylene fabric, it offers a robust barrier against particles, microorganisms, and non-hazardous liquids. The hood is processed and individually packaged in an ISO Class 4 cleanroom to minimize particle shedding and ensure high garment cleanliness.
  • Provides an excellent barrier to particles, microorganisms, and non-hazardous liquids
  • Suitable for ISO Classes 4-9 cleanrooms
  • Constructed with bound seams for enhanced protection
  • Treated with a topical antistatic agent
  • Gamma irradiated to achieve a sterility assurance level of 10-6
  • Free of natural rubber latex
  • Individually packaged for convenience in controlled settings