Learn More
AdipoGen Isocarbostyril
SDP
Supplier:  AdipoGen CDXI0197G025
Encompass_Preferred
Chemical. CAS 491-30-5. Formula C9H7NO. MW 145.16. Building block used for the synthesis of pharmacophores and other types of agents.
Catalog No. 50-256-0631