Learn More
Cayman Chemical Isocarboxazid, Purity: ≥98%, CAS: 59-63-2, 10 mg
SDP
Catalog No. 502994163
Encompass_Preferred
An inhibitor of MAO (IC50 = 4.8 μM for rat brain MAO); induces a 4-fold increase in tryptamine action in isolated rat fundal strips at a concentration of 50 nM; potentiates tryptamine toxicity (LD50 = 8 mg/kg following subcutaneous administration of 250 mg/kg tryptamine; inhibits 90% of MAO activity in isolated rat hearts and reduces isoproterenol-induced cardiomegaly in rats; increases levels of dopamine and norepinephrine and reduces levels of the monoamine metabolites DOPAC, HVA, and 5-HIAA in mouse brain
Catalog No. 50-299-4163 Supplier Cayman Chemical Supplier No. 2362510 MG
An inhibitor of MAO (IC50 = 4.8 μM for rat brain MAO); induces a 4-fold increase in tryptamine action in isolated rat fundal strips at a concentration of 50 nM; potentiates tryptamine toxicity (LD50 = 8 mg/kg following subcutaneous administration of 250 mg/kg tryptamine; inhibits 90% of MAO activity in isolated rat hearts and reduces isoproterenol-induced cardiomegaly in rats; increases levels of dopamine and norepinephrine and reduces levels of the monoamine metabolites DOPAC, HVA, and 5-HIAA in mouse brain