Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isocitric acid | 320-77-4 | MFCD00065926 | 192.12 g/mol | C6H8O7 | 5 MG
SDP

Catalog No. 5000264149
Encompass_Preferred

Isocitric acid (Standard) is an analytical reference standard intended for research and analytical applications. The material corresponds to isocitric acid (CAS 320-77-4) with molecular formula C6H8O7 and molecular weight 192.12 g/mol. Users should refer to the certificate of analysis for exact purity and recommended storage conditions.

  • Analytical reference standard for method development and calibration.
  • Confirmed identity with CAS 320-77-4 and formula C6H8O7.
  • Available in small pack sizes suitable for analytical use, including 5 mg.
  • Certificate of analysis documents purity and storage recommendations.

Catalog No. 50-002-64149 Supplier Medchemexpress LLC Supplier No. HY113228R5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isocitric acid (Standard) is an analytical reference standard intended for research and analytical applications. The material corresponds to isocitric acid (CAS 320-77-4) with molecular formula C6H8O7 and molecular weight 192.12 g/mol. Users should refer to the certificate of analysis for exact purity and recommended storage conditions.

  • Analytical reference standard for method development and calibration.
  • Confirmed identity with CAS 320-77-4 and formula C6H8O7.
  • Available in small pack sizes suitable for analytical use, including 5 mg.
  • Certificate of analysis documents purity and storage recommendations.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.