Learn More
Medchemexpress LLC Isocitric acid (standard) | 320-77-4 | 99.9% | 192.12 | 1 MG
SDP
Catalog No. 5000437250
Encompass_Preferred
Isocitric acid (Standard) is the analytical standard of Isocitric acid. This product is intended for research and analytical applications. It is an endogenous metabolite found in saliva and cellular cytoplasm that can be used for research related to Alzheimer's Disease, Lewy Body Dementia, and Anoxia. This compound is an analytical standard used as a reference for assays.
  • Commonly employed in qualitative, quantitative, and methodological research experiments.
  • Used in HPLC, GC, and MS.
Catalog No. 50-004-37250 Supplier Medchemexpress LLC Supplier No. HY113228R1MG
Isocitric acid (Standard) is the analytical standard of Isocitric acid. This product is intended for research and analytical applications. It is an endogenous metabolite found in saliva and cellular cytoplasm that can be used for research related to Alzheimer's Disease, Lewy Body Dementia, and Anoxia. This compound is an analytical standard used as a reference for assays.
  • Commonly employed in qualitative, quantitative, and methodological research experiments.
  • Used in HPLC, GC, and MS.