Learn More
Medchemexpress LLC Isocitric acid (Standard) | 320-77-4 | 99.91% | 192.12 | 10 MG
SDP
Catalog No. 5000437843
Encompass_Preferred
Isocitric acid (Standard) is the analytical standard of Isocitric acid (HY-113228). This product is intended for research and analytical applications. It is an endogenous metabolite present in Saliva and Cellular_Cytoplasm that can be used for the research of Alzheimer's Disease, Lewy Body Dementia and Anoxia.
  • Intended for research and analytical applications.
  • Endogenous metabolite present in saliva and cellular_cytoplasm.
  • Can be used for the research of Alzheimer's disease, Lewy body dementia and Anoxia.
  • The compound is the grade of analytical standard, which is the reference standard supplied assay.
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
Catalog No. 50-004-37843 Supplier Medchemexpress LLC Supplier No. HY113228R10MG
Isocitric acid (Standard) is the analytical standard of Isocitric acid (HY-113228). This product is intended for research and analytical applications. It is an endogenous metabolite present in Saliva and Cellular_Cytoplasm that can be used for the research of Alzheimer's Disease, Lewy Body Dementia and Anoxia.
  • Intended for research and analytical applications.
  • Endogenous metabolite present in saliva and cellular_cytoplasm.
  • Can be used for the research of Alzheimer's disease, Lewy body dementia and Anoxia.
  • The compound is the grade of analytical standard, which is the reference standard supplied assay.
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.