Learn More
Medchemexpress LLC Isocitric acid (Standard) | 320-77-4 | 99.91% | 192.12 | 100 MG
SDP
Catalog No. 5000435557
Encompass_Preferred
Isocitric acid (Standard) is the analytical standard of Isocitric acid (HY-113228). This product is intended for research and analytical applications. It is an endogenous metabolite present in saliva and cellular cytoplasm that can be used for the research of Alzheimer's Disease, Lewy Body Dementia, and Anoxia. The compound is the grade of analytical standard, which is the reference standard supplied assay.
  • Analytical standard for isocitric acid.
  • Intended for research and analytical applications.
  • Endogenous metabolite present in saliva and cellular cytoplasm.
  • Can be used for the research of Alzheimer's disease, Lewy body dementia and anoxia.
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
Catalog No. 50-004-35557 Supplier Medchemexpress LLC Supplier No. HY113228R100MG
Isocitric acid (Standard) is the analytical standard of Isocitric acid (HY-113228). This product is intended for research and analytical applications. It is an endogenous metabolite present in saliva and cellular cytoplasm that can be used for the research of Alzheimer's Disease, Lewy Body Dementia, and Anoxia. The compound is the grade of analytical standard, which is the reference standard supplied assay.
  • Analytical standard for isocitric acid.
  • Intended for research and analytical applications.
  • Endogenous metabolite present in saliva and cellular cytoplasm.
  • Can be used for the research of Alzheimer's disease, Lewy body dementia and anoxia.
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.