Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ IsoClean™ Cleanroom Front Snap Frocks, Bound Neck
GREENER_CHOICE

Catalog No. 190881124B Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Packaging Type:
Bulk Packaged
Individually Packaged
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Packaging Type
19-088-1124B Medium Bulk Packaged
19-0881125A Small Individually Packaged
19-088-1125B Medium Individually Packaged
19-088-1125C Large Individually Packaged
19-088-1125D X-Large Individually Packaged
19-088-1125E 2X-Large Individually Packaged
19-088-1125F 3X-Large Individually Packaged
19-088-1124A Small Bulk Packaged
19-088-1124C Large Bulk Packaged
19-088-1124D X-Large Bulk Packaged
19-088-1124E 2X-Large Bulk Packaged
19-088-1124F 3X-Large Bulk Packaged
17-988-085 4X-Large Bulk Packaged
19-088-1125G 4X-Large Individually Packaged
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
This item is not returnable. View return policy
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Catalog No. 19-088-1124B Supplier DuPont™ Supplier No. IC270BWHMD003000
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.

Tyvek™ IsoClean™ provides an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash. Clean processed versions available.

  • Frocks provide body-length jacket design with front snap closures for easy donning and doffing
  • Made of Tyvek™, a spunbonded olefin fabric that is lightweight and flexible
  • Processed in a certified ISO Class 4 (formerly Class 10) cleanroom, antistatic treated and packaged in durable, antistatic bags
  • Frocks are ISO Class 5 (formerly Class 100) compatible
  • Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration
  • Elastic opening for tighter fit at wrist
  • Bound neck for lower particle shedding
  • Adjustable snap at neck for better fit
  • Clean processed garments are individually packaged
  • Garments not clean processed are bulk packaged in double transparent poly liners
TRUSTED_SUSTAINABILITY

Specifications

Size Medium
Material Tyvek
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Snaps
Cuff Style Elastic
Collar Type Bound
Latex Free Yes
Size (English) Chest 32 to 36 in.
Size (Metric) Chest 81 to 91 cm
Sleeve Style Set-in Sleeve
Packaging Type Bulk Packaged
Length (Metric) 101.6 cm
Length (English) Sleeve 35.5 in.
Length (Metric) Sleeve 90.17 cm
Description Not Clean Processed
Cleanroom Class Class 100 (ISO 5)
Length (English) 40 in.
Product Type Controlled Environments Frock
Seam Type Bound
For Use With (Application) Cleanroom, Pharmaceutical, Biotechnology, Medical Device Manufacturing, and Laboratory Environments
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.