Learn More
Medchemexpress LLC Isoconazole (nitrate) | 24168-96-5 | 98.0% | 1 ML
SDP
Catalog No. 5000412609
Encompass_Preferred
Isoconazole nitrate is a broad-spectrum antimicrobial agent exhibiting highly effective antimycotic and gram-positive antibacterial activity. It has a rapid rate of absorption and low systemic exposure potential. This compound causes a rapid reduction of cellular adenosine triphosphate (ATP) and induces a blockade of cell division in vitro by affecting the synthesis and organization of the cell membrane, leading to necrosis and death of fungal cells at higher concentrations.
  • Broad-spectrum antimicrobial agent
  • Highly effective antimycotic and gram-positive antibacterial activity
  • Rapid absorption and low systemic exposure potential
  • Causes rapid reduction of cellular adenosine triphosphate (ATP)
  • Induces blockade of cell division by affecting cell membrane synthesis and organization
  • Induces necrosis and death of fungal cells at higher concentrations
Catalog No. 50-004-12609 Supplier Medchemexpress LLC Supplier No. HYB144410MM/1ML
Isoconazole nitrate is a broad-spectrum antimicrobial agent exhibiting highly effective antimycotic and gram-positive antibacterial activity. It has a rapid rate of absorption and low systemic exposure potential. This compound causes a rapid reduction of cellular adenosine triphosphate (ATP) and induces a blockade of cell division in vitro by affecting the synthesis and organization of the cell membrane, leading to necrosis and death of fungal cells at higher concentrations.
  • Broad-spectrum antimicrobial agent
  • Highly effective antimycotic and gram-positive antibacterial activity
  • Rapid absorption and low systemic exposure potential
  • Causes rapid reduction of cellular adenosine triphosphate (ATP)
  • Induces blockade of cell division by affecting cell membrane synthesis and organization
  • Induces necrosis and death of fungal cells at higher concentrations