Learn More
Medchemexpress LLC Isocorydine | 475-67-2 | MFCD00075905 | 100.0% | 341.40 | C20H23NO4 | 10 MG
SDP
Catalog No. 5000330268
Encompass_Preferred
Isocorydine is a naturally occurring aporphine alkaloid supplied as a high-purity research standard for biochemical and pharmacological studies. Reports indicate combination with doxorubicin shows potential activity against hepatocellular carcinoma.
  • High purity for reliable experimental results.
  • Characterized: CAS 475-67-2, formula C20H23NO4, mw 341.40.
  • Available as solid and as a 10 mM solution in DMSO.
  • Suitable for in vitro and in vivo preclinical research.
  • Certificate of analysis and safety data available.
Catalog No. 50-003-30268 Supplier Medchemexpress LLC Supplier No. HYN259110MG
Isocorydine is a naturally occurring aporphine alkaloid supplied as a high-purity research standard for biochemical and pharmacological studies. Reports indicate combination with doxorubicin shows potential activity against hepatocellular carcinoma.
  • High purity for reliable experimental results.
  • Characterized: CAS 475-67-2, formula C20H23NO4, mw 341.40.
  • Available as solid and as a 10 mM solution in DMSO.
  • Suitable for in vitro and in vivo preclinical research.
  • Certificate of analysis and safety data available.