Learn More
Medchemexpress LLC HY-N2591 5mg Medchemexpress, Isocorydine CAS:475-67-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN25915MG
Encompass_Preferred
Medchemexpress, HY-N2591 5mg Isocorydine CAS:475-67-2 Isocorydine is isolated from Dicranostigma leptopodum (Maxim.) Fedde (DLF). Isocorydine combines with Doxorubicin (DOX) has a promising potential to eradicate hepatocellular carcinoma (HCC). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0404