Learn More
Medchemexpress LLC (-)-Isocorypalmine (Tetrahydrocolumbamine) | 483-34-1 | 341.40 | 1 MG
SDP
Catalog No. 5000416802
Encompass_Preferred
(-)-Isocorypalmine (Tetrahydrocolumbamine), isolated from the crude base fraction of *Corydalis chaerophylla*, is a dopamine receptor ligand. Recombinant CYP719A21 displays strict substrate specificity and high affinity (Km=4.63 ± 0.71 μM) for (-)-Isocorypalmine.
  • Isolated from the crude base fraction of *Corydalis chaerophylla*
  • Dopamine receptor ligand
  • Recombinant CYP719A21 displays strict substrate specificity and high affinity
Catalog No. 50-004-16802 Supplier Medchemexpress LLC Supplier No. HYN09271MG
(-)-Isocorypalmine (Tetrahydrocolumbamine), isolated from the crude base fraction of *Corydalis chaerophylla*, is a dopamine receptor ligand. Recombinant CYP719A21 displays strict substrate specificity and high affinity (Km=4.63 ± 0.71 μM) for (-)-Isocorypalmine.
  • Isolated from the crude base fraction of *Corydalis chaerophylla*
  • Dopamine receptor ligand
  • Recombinant CYP719A21 displays strict substrate specificity and high affinity