Learn More
Cayman Chemical Isocurcumenol 10mg
SDP
Catalog No. 502990128
Encompass_Preferred
A sesquiterpene with diverse biological activities; inhibits LPS-induced production of NO in isolated mouse peritoneal macrophages; reduces LPS/D-galactosamine-induced increases in serum levels of GOT and GPT in a mouse model of liver injury at 50 mg/kg
Catalog No. 50-299-0128 Supplier Cayman Chemical Supplier No. 3071010 MG
A sesquiterpene with diverse biological activities; inhibits LPS-induced production of NO in isolated mouse peritoneal macrophages; reduces LPS/D-galactosamine-induced increases in serum levels of GOT and GPT in a mouse model of liver injury at 50 mg/kg